Adhesive Vinyl GHS OSHA Labels 'Phosphine', HCL Labels
Supplier: HCL Label
GH10297VN0023 GH10297VN0035 GH10297VN0047 GH20297VN0023 GH20297VN0035 GH20297VN0047 GH30297VN0023 GH30297VN0035 GH30297VN0047 GH40297VN0023 GH40297VN0035 GH40297VN0047
103020-470PK 74.03 USD InStock
103020-470 103020-472 103020-474 103020-476 103020-478 103020-480 103027-990 103027-992 103027-994 103027-996 103028-398 103028-400 103036-248 103036-250 103036-252 103036-254 103036-256 103036-258 103044-076 103044-078 103044-080 103044-082 103044-084 103044-086
Adhesive Vinyl GHS OSHA Labels 'Phosphine', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty