Adhesive Vinyl GHS OSHA Labels 'Hexafluoro-1 3-Butadiene', HCL Labels
Supplier: HCL Label
GH11294VN0023 GH11294VN0035 GH11294VN0047 GH21294VN0023 GH21294VN0035 GH21294VN0047 GH31294VN0023 GH31294VN0035 GH31294VN0047 GH41294VN0023 GH41294VN0035 GH41294VN0047
103024-302PK 74.03 USD InStock
103024-302 103024-304 103024-306 103024-308 103024-310 103024-312 103032-368 103032-370 103032-372 103032-374 103032-376 103032-378 103039-988 103039-990 103039-992 103039-994 103040-284 103040-286 103047-700 103047-702 103047-704 103047-706 103047-708 103047-710
Adhesive Vinyl GHS OSHA Labels 'Hexafluoro-1 3-Butadiene', HCL Labels
Labels
These ready-made laminated adhesive vinyl labels comply with the updated OSHA HazCom secondary container labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS). All sizes are protected with a chemical resistant lamination and a shelf/outdoor life of two years.
Certificaciones: RoHS compliant.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty