Adhesive Vinyl GHS OSHA Labels 'Potassium Iodide', HCL Labels
Supplier: HCL Label
GH1X840VN0023 GH1X840VN0035 GH1X840VN0047 GH2X840VN0023 GH2X840VN0035 GH2X840VN0047 GH3X840VN0023 GH3X840VN0035 GH3X840VN0047 GH4X840VN0023 GH4X840VN0035 GH4X840VN0047
103026-814PK 74.03 USD InStock
103026-814 103026-816 103026-818 103026-820 103026-822 103026-824 103035-560 103035-562 103035-564 103035-566 103035-568 103035-570 103041-972 103041-974 103041-976 103041-978 103041-980 103041-982 103049-786 103049-788 103049-790 103049-792 103050-698 103050-700
Adhesive Vinyl GHS OSHA Labels 'Potassium Iodide', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty