Mueller Hinton Agar, Plates, Northeast Lab Services
Supplier: HERAEUS SILICA & METALS LTD
P2000 P1460
101374-188PK 67.23 USD InStock
101374-188 101374-038
Mueller Hinton Agar, Plates, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Two fill volumes are available to meet different lab needs.
  • Double Bagged Formulations Available
  • Different Fill Volumes Available
  • Prepared Media
  • Size: 15 mm x 100 mm
  • Standard Petri Dish
NEL produces a wide variety of agar-based media in many sizes and types of bottles, both glass and autoclavable plastic.
NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.
Certificaciones: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty