Sabouraud Dextrose Agar, Plates, Northeast Lab Services
Supplier: HERAEUS SILICA & METALS LTD
P1550 P4025
101374-098PK 33.65 USD InStock
101374-098 101374-292
Sabouraud Dextrose Agar, Plates, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Agar is available in two media formulations and sizes.
  • Deep Fill Available
  • Prepared Medium
  • Standard Petri Dish Available
  • Lecithin and Polysorbate 80 Available
  • Fill Volume: 24 or 14 mL.
The prepared medium has a fill volume of 24 or 14 mL.
NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.
Certificaciones: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty