Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

UVP Pen-Ray® Zinc and Cadmium Lamps, Analytik Jena

Supplier: Analytik Jena US
90-0069-05 90-0069-06 90-0071-03 90-0071-06
89237-328EA 1020.96 USD InStock
89237-328 89237-330 89237-334 89237-336
UVP Pen-Ray® Zinc and Cadmium Lamps, Analytik Jena
Lights Spectrum Lamps
Zinc and cadmium lamps are designed for use with UV absorption cells and detectors and are often used in air pollution monitoring devices.

  • Cadmium peak lines are at 229 nm and 326 nm
  • Electrical leads are available in either side (radial) or end (axial) configurations

Zinc lamps can substitute for deuterium lamps due to the zinc lamps superior thermal stability, long life and prominent 214 nm energy peak.

Order Now
 

ORDER


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Description Length Supplier No. VWR Catalog Number Unit Price Quantity
Zinc Lamps, Radial, Cord length 203 mm 110 mm 90-0069-05 89237-328 Each
Retrieving
Product available on GSA Advantage®
Zinc Lamps, Axial, Cord length 203 mm 110 mm 90-0069-06 89237-330 Each
Retrieving
Product available on GSA Advantage®
Cadmium Lamps, Radial, Cord length 203 mm 110 mm 90-0071-03 89237-334 Each
Retrieving
Product available on GSA Advantage®
Cadmium Lamps, Axial, Cord length 203 mm 110 mm 90-0071-06 89237-336 Each
Retrieving
Product available on GSA Advantage®

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty