Blotting Papers, Advansta
Supplier: ADVANSTA INC MS
L-07045-060 L-07046-060
103254-744EA 58.85 USD InStock
103254-744 103254-746
Blotting Papers, Advansta
Blotting Paper
Pre-cut paper for electrophoretic transfers.
  • Conveniently pre-cut to work with most mini gel transfer assemblies
  • Standardized with uniform thickness for reproducibility
  • Tested to perform consistently without artifacts
  • Available sizes include 7x9cm and 8x10cm
Advansta's pre-cut Blotting Papers are ideal for use in transfer sandwiches and cassettes when proteins are transferred electrophoretically from a gel to a nitrocellulose, PVDF, or other membrane. The Advansta Blotting Papers are available in two sizes (7x9 or 8x10cm) to work with multiple transfer setups and membrane sizes.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty