Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Hydroxymethyl)-1-methylpyrrolidin-2-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

115640-500MG 115640-1G 115640-5G
102897-032EA 1867.69 USD InStock
102897-032 102897-034 102897-036
5-(Hydroxymethyl)-1-methylpyrrolidin-2-one ≥95%
5-(Hydroxymethyl)-1-methylpyrrolidin-2-one
Hazard: Irritant


Formula: C₆H₁₁NO₂
MDL Number: MFCD17010015
CAS Number: 89531-41-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty