Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Arabinose ≥99% (by HPLC), Calbiochem®

Supplier: EMD MILLIPORE (BIOSCIENCES)

178680-25GM 178680-100GM
80502-854EA 96.68 USD InStock
80502-854 80502-852
L(+)-Arabinose ≥99% (by HPLC), Calbiochem®
L(+)-Arabinose
A pectin sugar widely distributed in plants in the form of complex polysaccharides.

Special Instructions: Following reconstitution, filter-sterilize and store at room temperature (20 °C). Stock solutions are stable for up to six months at room temperature.


Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Form White solid
Purity ≥99%
Storage 15 to 30 °C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty