Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD5 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 53-7.3]

Supplier: Tonbo Biosciences
35-0051-U025 35-0051-U100
75875-868EA 45.1 USD InStock
75875-868 75875-870
Anti-CD5 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 53-7.3]
Antibodies
The 53-7.3 monoclonal antibody is specific for mouse CD5, a 67 kDa protein that is expressed on most thymocytes, mature T cells and the B-1 subset of B cells. CD5 is a member of the scavenger receptor cysteine-rich protein superfamily and is a ligand for the c-type lectin CD72. CD5 is also known as Leu-1 and functions to modulate antigen receptor signaling on T and B cells.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty