BCG-Dextrose HiVeg™ Agar, Snyder Test, Himedia
Supplier: PAESEL+LOREI GMBH&CO
MV106-500G MV106-2.5KG MV106-5KG
61000-428EA 281.11 USD InStock
61000-428 89227-560 89227-570
BCG-Dextrose HiVeg™ Agar, Snyder Test, Himedia
Media for Microbiology Dehydrated Media for Microbiology
Used for the estimation of Lactobacilli counts as an indication of caries activity.
  • Prepared by using HiVeg peptone in place of Peptic digest of animal tissue
  • Free of BSE/TSE risks
  • Modification of Snyder Test Agar
B.C.G. Dextrose HiVeg Agar is the modification of Snyder Test Agar, which was formulated by Snyder to estimate relative numbers of Lactobacilli in saliva and for diagnosis of caries activity.
Dextrose HiVeg Agar is prepared by using HiVeg peptone in place of Peptic digest of animal tissue, which makes the medium free of BSE/TSE risks.
Precaución: Store Dry Medium - below 30 °C, Prepared Medium 2 - 8 °C.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty