GeneBlaster™ Transfection Boosters, OZ Biosciences
Supplier: Med-Lab-Service
GB20010 GB20011 GB20012 GB20013 GB20014
103258-166EA 205.22 USD InStock
103258-166 103258-168 103258-170 103258-326 103258-328
GeneBlaster™ Transfection Boosters, OZ Biosciences
Transfection Reagents
To optimize your transfection experiments, OZ Biosciences has created GeneBlasters Transfection Boosters.
  • Higher gene expression in many cells
  • Convenient for a large panel of adherent and suspension cells
  • Prolong in vitro gene expression
  • Successful with all genetic vectors
  • Simple, rapid, and easy-to-use
  • Economical
GeneBlasters Transfection Boosters is an innovative and efficient solution to improve gene expression levels. GeneBlaster kits are a set of chemicals designed to get higher and longer transgene expression levels.
Información de suministro: Shipped at room temperature
Envase: Store all Geneblasters at +4 °C except GeneBlaster Emerald at -20 °C
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty