Microplate Sealing Mats
Supplier: Eppendorf
0030127960 0030127978
75874-653PK 321.88 USD InStock
75874-653 75874-655
Microplate Sealing Mats
Seals Microplate Seals
Secure your samples with confidence using Eppendorf® Sealing mats, the ideal solution for incubation and short-term storage needs. These reusable mats can be autoclaved and come in a 'PCR Clean' purity grade (free of RNase, DNase, Human DNA and PCR inhibitors).
  • Sealing mats are reusables and can be autoclaved
  • Suitable for short-term storage
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty