Polypropylene Carboys, with Handles, Chemglass
Supplier: Chemglass
CG-871-10 CG-871-20
10034-154EA 203.85 USD InStock
10034-154 10034-470
Polypropylene Carboys, with Handles, Chemglass
Bottles Carboys
Carboys are made of white autoclavable medical grade polypropylene resin, and can be used for storing and transporting large amounts of culture media, distilled water, and other solutions.
  • Wide handles for ease of carrying and pouring
  • Comes with molded-in graduations
  • Complete with white 83 mm leak-proof polypropylene closure
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty