Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hydroxide ∼45% in aqueous solution for HPLC, Fluka™

Supplier: BURDICK & JACKSON FKA SIGMA
Danger

03564-100ML 03564-500ML
BJ03564-100MLEA 76 USD InStock
BJ03564-100ML BJ03564-500ML
Potassium hydroxide ∼45% in aqueous solution for HPLC, Fluka™
Potassium hydroxide


Formula: KOH
Boiling Pt: 100 °C
Melting Pt: –6 °C
Density: 1.140 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II
Order Now
Specification Test Results
Assay 40.5 - 49.5 %
Assay of K₂CO₃ Max. 0.3 %
Suitability test (HPLC) complying
Density (D 20/4) 1.440 - 1.480
Absorbance at 500 nm Max. 0.02
Absorbance at 310 nm Max. 0.02
Absorbance at 300 nm Max. 0.03
Absorbance at 290 nm Max. 0.05
Absorbance at 240 nm Max. 0.1

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty