Chrome Steel Bar Magnet
Supplier: American Scientific
3101-33-B 3101-33
470225-330EA 26.6 USD InStock
470225-330 470225-382
Chrome Steel Bar Magnet
Magnets
For demonstrations, plotting magnetic fields.
  • Sturdy zinc plated magnets
  • Comes in marked and unmarked styles
Bar Magnet, made from magnetic steel strips, zinc plated. Supplied in pairs with cardboard separator and plated soft iron keepers.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty