Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dimethoxytrityl chloride ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

D2181-100GM D2181-25GM D2181-5GM
10860-454EA 771.95 USD InStock
10860-454 10860-456 10860-458
4,4'-Dimethoxytrityl chloride ≥98%
4,4'-Dimethoxytrityl chloride
Storage temperature - HYGROSCOPIC: Keep tightly closed.

Precaución: Storage temperature - HYGROSCOPIC: Keep tightly closed.


Formula: C₂₁H₁₉ClO₂
MW: 338.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008409
CAS Number: 40615-36-9
Order Now
Specification Test Results
Assay Approx. 98 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty