Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-nitrobenzoic acid ≥98.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

A1892-1GM A1892-5GM A1892-25GM
10860-318EA 50.9 USD InStock
10860-318 10860-320 10860-322
5-Amino-2-nitrobenzoic acid ≥98.0%
5-Amino-2-nitrobenzoic acid
GHS hazard : Exclamation mark


Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Boiling Pt: 481
Melting Pt: 234
Storage Temperature: Ambient
MDL Number: MFCD00035748
CAS Number: 13280-60-9
Order Now
Specification Test Results
Assay (Titration) 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty