Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Acetylamino)hexanoic acid ≥98.5%

Supplier: SPECTRUM CHEMICAL MFG CORP

A1042-25GM A1042-5GM A1042-100GM
700013-316EA 291.7 USD InStock
700013-316 700013-318 700013-314
6-(Acetylamino)hexanoic acid ≥98.5%
6-(Acetylamino)hexanoic acid
6-Acetamidocaproic Acid is an unnatural amino acid derivative that finds utility in peptide synthesis.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004424
CAS Number: 57-08-9
Order Now
Specification Test Results
Assay 98.5% min.
Melting Point As Reported

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty