Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-n-Hexylcinnamaldehyde ≥85%

Supplier: SPECTRUM CHEMICAL MFG CORP

H2224-250ML H2224-1LT
700003-784EA 272.7 USD InStock
700003-784 700003-782
α-n-Hexylcinnamaldehyde ≥85%
α-n-Hexylcinnamaldehyde
GHS hazard : Exclamation mark


Formula: C₁₅H₂₀O
MW: 216.3187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006989
CAS Number: 101-86-0
Order Now
Specification Test Results
Assay Approx. 85 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty