Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Terpinyl acetate ≥97.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

T2428-500ML T2428-100ML
700004-640EA 293.05 USD InStock
700004-640 700004-638
Terpinyl acetate ≥97.0%
Terpinyl acetate
alpha-Terpinyl Acetate is a perfumery and flavoring ingredient.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₂H₂₀O₂
Density: 0.953
Storage Temperature: Ambient
MDL Number: MFCD00037155
CAS Number: 80-26-2
Order Now
Specification Test Results
Assay 97.0 % min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty