Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Humic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

H1452-500GM H1452-100GM
700009-820EA 411.84 USD InStock
700009-820 700009-816
Humic acid
Humic acid


Formula: C₉H₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00147177
CAS Number: 1415-93-6
Order Now
Specification Test Results
Appearance Dark Brown, Powd

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty