Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxyethyl cellulose NF

Supplier: SPECTRUM CHEMICAL MFG CORP

H1236-500GM H1236-2.5KG H1236-45KGBL H1236-100GM H1236-12KGBL
95033-576EA 351.8 USD InStock
95033-576 95033-574 700013-008 95033-572 75811-356
Hydroxyethyl cellulose NF
Hydroxyethyl cellulose


Storage Temperature: Ambient
MDL Number: MFCD00072770
CAS Number: 9004-62-0
Order Now
Specification Test Results
Viscosity of 1% aqueous solution @25°C 2500 - 7500 cP
pH (1%) 6.0 - 8.5
Loss On Drying max. 10.0%
Residue On Ignition max. 5.0%
Lead (Pb) max. 0.001%

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty