Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium linoleate ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP

L3096-5GM L3096-1GM
700005-466EA 620.05 USD InStock
700005-466 700005-464
Sodium linoleate ≥97%
Sodium linoleate
Linoleic Acid Sodium Salt, a polyunsaturated omega-6 fatty acid, is becoming increasingly popular in the personal care products industry because of its beneficial anti-inflammatory, acne reductive, and moisture retentive properties when applied topically on the skin.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₈H₃₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00063202
CAS Number: 822-17-3
Order Now
Specification Test Results
Assay Approx. 97 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty