Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Lysine-L-glutamate

Supplier: SPECTRUM CHEMICAL MFG CORP

L2091-100GM L2091-1KGBL L2091-5KGBL
700014-686EA 204.9 USD InStock
700014-686 700014-688 700014-690
L-Lysine-L-glutamate
L-Lysine-L-glutamate
L-Lysine L-Glutamate Salt is a glutamate salt of L-lysine. L-Lysine is an essential amino acid in human. It is metabolized to acetyl-CoA. It is widely used as an ingredient in infusion.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₁H₂₃N₃O₆
MW: 293.32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038974
CAS Number: 5408-52-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty