Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl-N-dodecylbetaine solution 35%

Supplier: SPECTRUM CHEMICAL MFG CORP

D1658-500GM D1658-25GM D1658-100GM
700014-150EA 498.7 USD InStock
700014-150 700014-148 700014-146
N,N-Dimethyl-N-dodecylbetaine solution 35%
N,N-Dimethyl-N-dodecylbetaine
GHS hazard : Skull and crossbones
UN2810


Formula: C₁₆H₃₃NO₂
Density: 1.03
Storage Temperature: Ambient
MDL Number: MFCD00084742
CAS Number: 683-10-3
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Assay 33.5 - 37.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty