Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nonoxynol-9

Supplier: SPECTRUM CHEMICAL MFG CORP

N1217-500MLG N1217-100ML N1217-200LTB
700014-894EA 522.75 USD InStock
700014-894 700014-890 700014-892
Nonoxynol-9
Nonoxynol-9
Nonoxynol 9 is also known as N-9 is used as a surfactant. It is an ingredient in several different types of cosmetic and cleaning products nonionic surfactants.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₃₃H₆₀O₁₀
Storage Temperature: Ambient
CAS Number: 26571-11-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty