Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium sulfite, Powder

Supplier: SPECTRUM CHEMICAL MFG CORP

P1609-2.5KG P1609-500GM
700001-468EA 820.05 USD InStock
700001-468 700001-470
Potassium sulfite, Powder
Potassium sulfite
Potassium Sulfite, Powder has been most often used as a food additive and preservative. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


Formula: K₂SO₃
Storage Temperature: Ambient
MDL Number: MFCD00011387
CAS Number: 10117-38-1
Order Now
Specification Test Results
Assay 88.5 - 112.5 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty