Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thyme oil, white

Supplier: SPECTRUM CHEMICAL MFG CORP

T1048-100KGB T1048-100ML T1048-500MLG
700010-834EA 9958.55 USD InStock
700010-834 700010-836 700010-838
Thyme oil, white
Thyme oil
Thyme Oil, White, an essential oil of thyme, has many applications including as an antiseptic and as an effective agent against certain fungi.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Density: 0.917
Storage Temperature: Ambient
MDL Number: MFCD00148640
CAS Number: 8007-46-3
UN: 2924
ADR: 38 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty