Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thymol iodide, powder, purified

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

TH180-125GM TH180-500GM TH180-25GM TH180-12KGBL
700001-316EA 632.25 USD InStock
700001-316 700001-324 700001-322 700001-318
Thymol iodide, powder, purified
Thymol iodide
Thymol Iodide, Powder, Purified is used in the manufacture of some polymers, lacquers, adhesives, waxes and polishes.

The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Precaución: Storage temperature - LIGHT SENSITIVE, REFRIGERATE: Keep tightly closed in light-resistant containers.


Formula: C₂₀H₂₄I₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00026409
CAS Number: 552-22-7
Order Now
Specification Test Results
Assay (I) 42.0 % min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty