Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trigonelline hydrochloride

Supplier: SPECTRUM CHEMICAL MFG CORP

T3921-5GM T3921-1GM
700009-594EA 529.1 USD InStock
700009-594 700009-592
Trigonelline hydrochloride
Trigonelline hydrochloride


Formula: C₇H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00077250
CAS Number: 6138-41-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty