Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylolpropane triacrylate, Technical Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

T1359-500GM T1359-25GM
700005-360EA 431.45 USD InStock
700005-360 700005-356 700005-358
Trimethylolpropane triacrylate, Technical Grade
Trimethylolpropane triacrylate
Trimethylolpropane Triacrylate, Technical, also known as TMPTA, is a trifunctional monomer used to manufacture adhesives, plastics, acrylic glue, and ink.

Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.


Formula: C₁₅H₂₀O₆
MW: 296.32 g/mol
Density: 1.10
Storage Temperature: Ambient
MDL Number: MFCD00008628
CAS Number: 15625-89-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty