Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-N-octanoyl-D-glucamine (MEGA-8), Ultrapure

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CC1-0007-G001 AG-CC1-0007-G005
102987-838EA 159.23 USD InStock
102987-838 102987-840
N-Methyl-N-octanoyl-D-glucamine (MEGA-8), Ultrapure
N-Methyl-N-octanoyl-D-glucamine (MEGA-8)
Non-ionic water-soluble detergent with nondenaturing properties. Transparent in the UV region. Relatively high critical micelle concentration (CMC) value (79mM/l). Ideal for studying the conformation and function of proteins, keeping the secondary and tertiary structure intact, therefore suited for solubilizing membrane proteins without altering biological activity. Efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins. Ideal for use as membrane protein solubilizer.


Formula: C₁₅H₃₁NO₆
MW: 321.4 g/mol
MDL Number: MFCD00134152
CAS Number: 85316-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty