Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Simvastatin

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0052-M010 AG-CN2-0052-M050
102987-854EA 34.62 USD InStock
102987-854 102987-856
Simvastatin
Simvastatin
Synthetic analog of Lovastatin. Potent competitive HMG-CoA reductase inhibitor. Anti-hypercholesterolemic agent. Cholesterol/isoprenoid biosynthesis inhibitor. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. Inhibits the isoprenylation of Ras and Rho family GTPases. Smooth muscle cell proliferation inhibitor. Anti-adhesive, immunomodulatory and anti-inflammatory compound. Proteasome modulator. Stimulates bone formation. Apoptosis inducer. Anticancer compound. Increases cellular resistance to anticancer agents such as doxorubicin and etoposides. Suppresses ICAM-1-LFA-1 interactions, which blocks virus replication and infection. Anti-hypertensive agent. Suppresses TNF-induced NF-kappaB activation.


Formula: C25H38O5
MDL Number: MFCD00072007
CAS Number: 79902-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty