Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ponasterone A

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0053-M001 AG-CN2-0053-M005 AG-CN2-0053-M025
102987-858EA 83.08 USD InStock
102987-858 102987-860 102987-862
Ponasterone A
Ponasterone A
A member of the ecdysteroid family. Ecdysone receptor (EcR) agonist. Analog of ecdysone with similar properties to muristerone A. Functional, reliable and economical substitute for muristerone A as an inducer for the ecdysone-inducible mammalian expression system. Induces expression of beta-galactosidase to levels similar to those obtained with muristerone A induction. Potent compound for ecdysteroid activity. Insect steroid hormone involved in regulating metamorphosis.


Formula: C₂₇H₄₄O₆
MW: 464.643 g/mol
CAS Number: 13408-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty