Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Α-Ecdysone

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0071-M001 AG-CN2-0071-M005 AG-CN2-0071-M010
102987-232EA 62.31 USD InStock
102987-232 102987-234 102987-236
Α-Ecdysone
Α-Ecdysone
Steroidal prohormone of the major insect molting hormone 20-hydroxyecdysone. A member of the ecdysteroid family. Ecdysone receptor (EcR) agonist. Induces the expression of genes coding for proteins that the larva requires, and it causes chromosome puffs (sites of high expression) to form in polytene chromosomes. Plays a key role in insect development, cell proliferaton, growth and apoptosis by controlling gene expression involved in moulting and metamorphosis. It acts through a heterodimeric receptor comprising the ecdysone receptor and the ultraspiracle proteins (USP). Appears in many plants mostly as a protection agent (toxins or antifeedants) against herbivorous insects. Used for controlled gene expression in scientific research, agriculture and medicine. Used for the development of selective insect growth regulators for use as environmentally benign insecticides.


Formula: C₂₇H₄₄O₆
MDL Number: MFCD00042683
CAS Number: 3604-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty