Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Cytisine

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0075-M005 AG-CN2-0075-M025
102987-246EA 27.69 USD InStock
102987-246 102987-248
(-)-Cytisine
(-)-Cytisine
Pyridine-like alkaloid. Nicotine agonist. Potent agonist at alpha3beta4 and alpha7 nicotinic acetylcholine receptors and partial agonist at alpha4beta2 nicotinic acetylcholine receptors. Shows analgesic, antihypertensive and inotropic activities. Used for smoking cessation. Might be used for binge-like ethanol drinking cessation. Iron-chelator and hydroxyl radical scavanger. Has antidepressant properties. Used in forensics in novel surface molecular imprinting techniques.


Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
MDL Number: MFCD00136048
CAS Number: 485-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty