Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiocolchicoside

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0076-M001 AG-CN2-0076-M005 AG-CN2-0076-M025
102987-250EA 48.46 USD InStock
102987-250 102987-252 102987-254
Thiocolchicoside
Thiocolchicoside
Potent competitive gamma-aminobutyric acid type A (GABAA) receptor antagonist and glycine receptor agonist. Weak nicotinic acetylcholine receptor agonist. Muscle relaxant. Anti-inflammatory. Has analgesic properties. Shows strong epileptogenic and convulsant activity. Anticancer compound through inhibition of NF-kappaB and NF-kappaB-regulated gene products Apoptosis inducer. Suppressed osteoclastogenesis induced by RANKL and tumor cells via the NF-kappaB signaling pathway. Therapeutic option for the management of bone metastatic disease.


Formula: C₂₇H₃₃NO₁₀S
MDL Number: MFCD07995056
CAS Number: 602-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty