Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

paxilline

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0167-M001 AG-CN2-0167-M005
102988-126EA 69.23 USD InStock
102988-126 102988-128
paxilline
Paxilline
Tremorgenic. Potent, selective and reversible inhibitor of high-conductance calcium-activated potassium (BKCa) channels. Reversible inhibitor of inositol 1,4,5-trisphosphate receptor (InsP(3)). Sarco/endoplasmic reticulum Ca2+ ATPase (SERCA) inhibitor. Liver X receptor (LXRalpha and LXRbeta; NR1H3 and NR1H2) agonist in biochemical and in vitro cell-based assays. Anticonvulsant. Apoptosis enhancer. Can sensitize various glioma cells to TRAIL-mediated apoptosis. Shown to protect neuronal cells against glutamate-induced cell death.


Formula: C₂₇H₃₃NO₄
MDL Number: MFCD00083464
CAS Number: 57186-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty