Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Asiatic acid

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0400-M005 AG-CN2-0400-M025
102988-146EA 55.38 USD InStock
102988-146 102988-148
Asiatic acid
Asiatic acid
Apoptosis inducer. Cell cycle arrest inducer. Anticancer compound Antioxidant. Hepatoprotective. Inhibits TGF-beta/Smad-mediated fibrogenesis. Stimulates wound healing. Anti-diabetic. Glycogen phosphorylase inhibitor. Neuroprotective. Modulates multiple targets associated with amyloid-beta precursor protein processing and amyloid-beta protein clearance. Down regulates BACE1 and increases ADAM10 maturation. Anti-hyperglycemic compound. Anti-inflammatory and antinociceptive compound. Antiangiogenic. Acetylcholinesterase (AChE) inhibitor. PPARgamma inhibitor through a C/EBPbeta-independent mechanisms. Anti-osteoporotic. Inhibits adipogenic differentiation of bone marrow stromal cells (BMSC).


Formula: C₃₀H₄₈O₅
MW: 488.709 g/mol
MDL Number: MFCD00238541
CAS Number: 464-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty