Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ursodeoxycholic acid

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0411-G001 AG-CN2-0411-G005
102988-172EA 55.38 USD InStock
102988-172 102988-174
Ursodeoxycholic acid
Ursodeoxycholic acid
Endogenous hydrophilic bile acid. Antioxidant. Cytoprotective against oxidative stress and cell death. Hepatoprotective at cellular and molecular level, including stabilization of membranes. Protects hepatocytes against bile acid-induced apoptosis. Antiapoptotic and antinecrotic. Targets the mitochondrial function and integrity, reduction of endoplasmatic stress and interactions with survival signals in cAMP, Akt, NF-kappaB, MAPK and PI3K signaling pathways. Modulator and finetuner of the p53-Mdm-2 complex. Chemopreventive against colorectal cancer by countering the tumor-promoting effects of secondary bile acids. Shows also effects on epidermal growth factor receptor (EGFR) signaling and COX-2 expression. Immunomodulator and anti-inflammatory compound. Modifies TLR4 and TLR9 signaling pathways and downregulates the production of proinflammatory tumor necrosis factor-alpha (TNF-alpha). Pregnane X receptor agonist. Neuroprotective. Inhibits neuronal apoptosis. Glucocorticoid Receptor (GR) agonist. Anticholestatic agent. Used to reduce cholesterol absorption and for cholesterol gallstone dissolution. Used to treat primary biliary cirrhosis (PBC). Interferes with the progression of non-alcoholic fatty liver disease (NAFLD)/NASH. Reduces CXCR3 expression. TIMP-1 inducer. ADAM17 inhibitor.


Formula: C₂₄H₄₀O₄
MW: 392.58
Boiling Pt: 547
Melting Pt: 205
MDL Number: MFCD00003680
CAS Number: 128-13-2
Merck Index: 12,10026
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty