Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Betulinic acid

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0417-M100 AG-CN2-0417-M500 AG-CN2-0417-G001
102988-198EA 83.08 USD InStock
102988-198 102988-200 102988-202
Betulinic acid
Betulinic acid
Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.


Formula: C₃₀H₄₈O₃
MW: 456.71 g/mol
MDL Number: MFCD00009619
CAS Number: 472-15-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty