Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Artemisinin

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0467-M100 AG-CN2-0467-M500 AG-CN2-0467-G001
102988-374EA 48.46 USD InStock
102988-374 102988-376 102988-378
Artemisinin
Artemisinin
Anti-malaria. Apoptosis inducer. Anti-cancer. Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO).


Formula: C₁₅H₂₂O₅
MW: 282.34 g/mol
MDL Number: MFCD00081057
CAS Number: 63968-64-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty