Amastatin Hydrochloride
Supplier: Van Der Voort Lucht en Milieu Amste
AG-CP3-7003-M001 AG-CP3-7003-M005
102988-528EA 62.31 USD InStock
102988-528 102988-530
Amastatin Hydrochloride
Proteins and Peptides
Slow, tight binding and competitive aminopeptidase (AP) inhibitor. Inhibits cytosolic leucine aminopeptidase, microsomal aminopeptidase M and bacterial leucine aminopeptidase, human serum aminopeptidase A (AP-A), aminopeptidase N (AP-N), tyrosine aminopeptidase, but not aminopeptidase B (AP-B).
Amastatin is without effect on trypsin, papain, chymotrypsin, elastase, pepsin or thermolysin. Inhibits completely the Suc-Ala-Ala-Pro-Leu-pNA amidolytic enzyme. Slightly inhibits the formation of angiotensin III (Ang III) from Angiotensin II through AP-A, but significantly increases the potency of angiotensin III and-angiotensin I. Moderate inhibitor of mitochondrial intermediate peptidase (MIP). Weak inhibitor of simian immunodeficiency virus protease (SIV-PR).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty