Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chelerythrine chloride

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CR1-0071-M001 AG-CR1-0071-M005 AG-CR1-0071-M025
102988-534EA 34.62 USD InStock
102988-534 102988-536 102988-538
Chelerythrine chloride
Chelerythrine chloride
Cell permeable potent inhibitor of protein kinase C. Does not inhibit tyrosine protein kinases, cAMP-dependent protein kinase or calcium/calmodulin-dependent protein kinase. Antiplatelet, anti-inflammatory, antibacterial and antitumor compound. Apoptosis inducer in cancer cells in vitro and in vivo. Activates MAPK and JUNK signaling pathways. Affects translocation of PKC from cytosol to plasma membrane. Neurite outgrowth stimulator. Inhibits binding of BclXL to Bak (IC50 = 1.5 µM) or Bad proteins and stimulates apoptosis in several cancer cell lines. Blocks human P2X7 receptor. Induces cell cycle arrest in G1 phase. Specific cyclooxygenase-2 inhibitor.


Formula: C₂₁H₁₈ClNO₄
MDL Number: MFCD00060717
CAS Number: 3895-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty