Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ajmalicine

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-A0214-M010 CDX-A0214-M250 CDX-A0214-GG25
102989-278EA 70.62 USD InStock
102989-278 102989-280 102989-282
Ajmalicine
Ajmalicine
Alkaloid used to study its effects as an antagonist of adrenergic and nicotinic receptors.


Formula: C₂₁H₂₄N₂O₃
MW: 352.43 g/mol
MDL Number: MFCD00042748
CAS Number: 483-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty