Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Penta-2,4-dienoic acid ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-B0002-G005 CDX-B0002-G025
102989-302EA 181.38 USD InStock
102989-302 102989-304
Penta-2,4-dienoic acid ≥97%
Penta-2,4-dienoic acid
Building block for synthesis.


Formula: C5H6O2
MW: 98.0999 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00014017
CAS Number: 626-99-3
Order Now
Specification Test Results
Source/Host Synthetic
Purity 97% (T)
Appearance/Color Light yellow crystalline.
Solubility Soluble in 1M sodium hydroxide.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty