Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥98% for dye

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-B0029-M500 CDX-B0029-G005
102989-322EA 416.77 USD InStock
102989-322 102989-324
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥98% for dye
bisBenzimide H 33258 trihydrochloride (Hoechst 33258)
Useful for staining DNA, chromosomes and nuclei. Binds to the minor groove of DNA at AT-rich sequences. This dye is commonly used for determining the DNA content of viable cells without detergent treatment or fixation and for fluorescence microscopy or flow cytometry.


Formula: C₂₅H₂₄N₆O.3HCl
MW: 533.88 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012679
CAS Number: 23491-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty