Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2-pyridylmethyl)amine

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-B0134-GG25 CDX-B0134-G005 CDX-B0134-G025
102989-382EA 70.62 USD InStock
102989-382 102989-384 102989-386
Bis(2-pyridylmethyl)amine
Bis(2-pyridylmethyl)amine
Building block for synthesis. DPA is a secondary amine with two picolyl substituents. The compound is a tridentate ligand in coordination chemistry and commonly used to produce Zn-based chemosensors/probes, such as Zinpry. As a tridentate ligand this compound provides three nitrogen donors that affords good selectivity for Zn2+ over biologically relevant metals such as Na+, K+, Mg2+ and Ca2+, and leaves coordination sites free for anion binding. The amino nitrogen of the DPA group is a good candidate as an electron donor in either photoinduced electron transfer or photoinduced charge transfer (PET or PCT) sensors. Zn(II)–DPA complexes are widely used in anion recognition and sensing.


Formula: C₁₂H₁₃N₃
MW: 199.26 g/mol
MDL Number: MFCD00129044
CAS Number: 1539-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty