Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Berberine chloride

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-B0212-G010 CDX-B0212-G025 CDX-B0212-G050
102989-404EA 78.92 USD InStock
102989-404 102989-406 102989-408
Berberine chloride
Berberine chloride
Natural fluorescent dye, used in histology for staining heparin in mast cells. An isoquinoline alkaloid, which shows antibacterial, antifungal, anti-inflammatory, anti-oxidant, antidiabetic, antiviral and anticancer activity. It inhibits cell growth and induces apoptosis in a variety of cancer cell lines. Broad spectrum enzyme inhibitor, affects N-acetyltransferase, cyclooxygenase-2, and topoisomerase activities and gene/protein expression. Increases adiponectin expression, upregulates LDLR and insulin receptor. Anti-hyperglycemic, by inhibition of aldose reductase, inducing glycolysis, preventing insulin resistance through increasing insulin receptor expression and acting like incretins. Activates AMPK and ERK. Potent, uncompetitive, reversible IDO 1 inhibitor with IC50 values of 7 µM (cell-based assay) and 9.3 µM (enzymatic assay), and Ki value 8µM.


Formula: C₂₀H₁₈ClNO₄
MDL Number: MFCD00011939
CAS Number: 633-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty